Learn More
Ohaus™ MB120 Moisture Analyzer
Artikelnummer. 15535029 Shop alle Ohaus produkter
Skift visning
Klik for at se tilgængelige muligheder
Connection Type:
USB host x 2USB deviceRS232
Pakningsstørrelse:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Connection Type Pakningsstørrelse
15535029 USB host x 2USB deviceRS232 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15535029 Leverandør Ohaus™ Leverandørnr. 30241169
  • ONLINE EXCLUSIVE PRICE
    62120.00 NOK
    46571.77 NOK / Pack of 1
    Save 15548.23 NOK
    25% Off
Se alle kampagner
In Stock
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
Measures moisture content with increased precision and efficiency.
  • Easily analyze specific material samples and determine the optimum drying temperature
  • 100 methods library makes testing faster
  • Store up to 1000 results for all methods for statistical analysis
  • Set up and lock temperature settings and methods parameters with admin access
  • Features wide, bright touchscreen display and intuitive menu navigation for fast, accurate moisture analysis
  • Fast infrared halogen heating technology
  • Uniform drying for precise, repeatable results
  • Flexible and powerful
  • Four drying profiles to ensure accuracy: Fast, Standard, Ramp, Step
  • Shut-Off criteria for automatic test completion
  • Features pan handler and removable parts for convenient maintenance
Certificeringer
CE, UL listed, CSA approved and FCC approved
TRUSTED_SUSTAINABILITY
Tekniske data
Model MB120
Outputs RS232, USB device, USB host
Repeatability 0.05% (3g sample), 0.015% (10g sample)
Width (Metric) Pan 90 mm
Dimensions (L x W x H) 355.00 x 211.00 x 180.000 mm
Electrical Requirements 120/240 VAC 50/60 Hz
Readability 0.01%/0.001 g
Display Type Full-Color 4.3 in Touchscreen WQVGA
Connection Type USB host x 2USB deviceRS232
Depth (Metric) 355.00 mm
Description Moisture Analyzer, MB120
Frequency 50/60 Hz
Height (English) 7.09 in.
Height (Metric) 180.000 mm
Humidity Range Moisture: 0.01% to 100%
Length (English) 13.98 in.
Length (Metric) 355.00 mm
Net Weight (English) 11.530 lb.
Net Weight (Metric) 5.230 kg
Sample Size 0.500 g to 120.000 g
Temperature Range (Metric) 40°C to 230°C
Voltage 120 VAC or 240 VAC
Width (English) 8.31 in.
Width (Metric) 211.00 mm
Vis mere Vis mindre
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.