Learn More
MedChemExpress Matrine
Description
Matrine has a variety of pharmacological effects, including anti-cancer, anti-oxidative stress, anti-inflammation and anti-apoptosis effects. Matrine is potential in the research of disease like human non-small cell lung cancer, hepatoma, papillary thyroid cancer and acute kidney injury (AKI).
Specifications
Specifications
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Molecular Weight (g/mol) | 248.36 g/mol |
| Shipping Condition | Room temperature |
| For Use With (Application) | Cancer-programmed cell death |
| CAS | 519-02-8 |
| Product Type | Natural Products |
| Purity | 98.0% |
| Solubility | DMSO : ≥ 50 mg/mL (201.32 mM) ∣H2O : 20 mg/mL (80.53 mM∣Need ultrasonic) |
| Synonym | Matridin-15-one∣ Vegard∣ α-Matrine |
| Target | Opioid Receptor∣Autophagy∣Mitophagy∣Ferroptosis∣Apoptosis |
| Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.