Learn More
Eppendorf™ Mastercycler™ X50
Product Code. 15886721 Shop All Eppendorf Products
Change view
Click to view available options
Description:
X50i
X50r
X50t
Content And Storage:
Aluminum Block and Pressure Lid
Silver Block
Max. Cooling Rate:
2.3°C/s
5°C/s
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Content And Storage Max. Cooling Rate Unit Size
15886721 X50t Aluminum Block and Pressure Lid 2.3°C/s Each
15856721 X50i Silver Block 5°C/s Each
15876721 X50r Aluminum Block and Pressure Lid 2.3°C/s Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15886721 Supplier Eppendorf™ Supplier No. 6306000010
  • ONLINE EXCLUSIVE PRICE
    120500.00 NOK
    106489.12 NOK / Pack of 1
    Save 14010.88 NOK
    11% Off
Estimated Shipment: 20-10-2026
Eppendorf™ Mastercycler™ X50 elegantly combined speed, flexibility and PCR optimization for research applications and standardization. A new highly intuitive touch screen concept puts all of those benefits always at your fingertips.
  • There are various blocks to choose from, from the fast silver block to the 384-well block
  • Touch screen control panel
  • Innovative 2D gradient for modern PCR optimization
  • X50 with touchscreen can control up to nine other devices (connected cyclers do not have a touch screen control panel)
  • Flexlid™ concept: Use of various consumables thanks to automatic lid height adjustment
  • Small footprint
  • Fast PCR
  • PCR optimization
  • Standard PCR
  • Cycle Sequencing
TRUSTED_SUSTAINABILITY
Specifications
Thermal Accuracy ±0.15°C
Content And Storage Aluminum Block and Pressure Lid
Voltage 110 to 230 V
Height (Metric) 33 cm
Width (Metric) 27.5 cm
Depth (Metric) 43 cm
Thermal Range 4°C to 99°C
Description X50t
Max. Cooling Rate 2.3°C/s