Learn More
Eppendorf™ Mastercycler™ X50
Product Code. 15856721 Shop All Eppendorf Products
Change view
Click to view available options
Description:
X50i
X50r
X50t
Content And Storage:
Aluminum Block and Pressure Lid
Silver Block
Max. Cooling Rate:
2.3°C/s
5°C/s
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Content And Storage Max. Cooling Rate Unit Size
15856721 X50i Silver Block 5°C/s Each
15876721 X50r Aluminum Block and Pressure Lid 2.3°C/s Each
15886721 X50t Aluminum Block and Pressure Lid 2.3°C/s Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15856721 Supplier Eppendorf™ Supplier No. 6301000012
  • ONLINE EXCLUSIVE PRICE
    112900.00 NOK
    99719.29 NOK / Pack of 1
    Save 13180.71 NOK
    11% Off
View all promotions
In Stock
Eppendorf™ Mastercycler™ X50 elegantly combined speed, flexibility and PCR optimization for research applications and standardization. A new highly intuitive touch screen concept puts all of those benefits always at your fingertips.
  • There are various blocks to choose from, from the fast silver block to the 384-well block
  • Touch screen control panel
  • Innovative 2D gradient for modern PCR optimization
  • X50 with touchscreen can control up to nine other devices (connected cyclers do not have a touch screen control panel)
  • Flexlid™ concept: Use of various consumables thanks to automatic lid height adjustment
  • Small footprint
  • Fast PCR
  • PCR optimization
  • Standard PCR
  • Cycle Sequencing
TRUSTED_SUSTAINABILITY
Specifications
Thermal Accuracy ±0.15°C
Content And Storage Silver Block
Voltage 110 to 230 V
Height (Metric) 33 cm
Width (Metric) 27.5 cm
Depth (Metric) 43 cm
Thermal Range 4°C to 99°C
Description X50i
Max. Cooling Rate 5°C/s