Learn More
Eppendorf™ Mastercycler Nexus SX1e, EU-Plug
Shop All Eppendorf Products
Click to view available options
:
Capacity: 96 x 0.2 mL PCR vessels, 1 PCR plate (96 wells)
Description
- Intuitive graphic programming
- Small footprint
- Universal block for ultimate flexibility
- Connect up to 2 Mastercycler nexus eco to your existing unit
- Available with gradient
- E-mail notification
- flexlid™ concept allows use of all types of consumables with automatic height adjustment of the lid
Specifications
Specifications
| Temperature Range (Metric) | 37 to 110°C (Lid), 4 to 99°C (Block) |
| Content And Storage | EU Plug |
| Dimensions | 25 × 41.2 × 32.1 cm |
| Capacity | 96 × 0.2 mL PCR Vessels, 1 PCR Plate (96-wells) |
| Electrical Requirements | 230 V, 50/60 Hz |
| For Use With (Application) | Standard PCR, Fast PCR, In situ PCR |
| User Interface | CAN_in/CAN_out, Ethernet, USB |
| No. of Programs | >700 |
| Weight | 10.4 kg |