Learn More
Eppendorf™ Mastercycler™ Nexus Gradient
Product Code. 12304943 Shop All Eppendorf Products
Change view
Click to view available options
Without GB Plug
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12304943 Without GB Plug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12304943 Supplier Eppendorf™ Supplier No. 6331000017
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Temperature Range (Metric) 30 to 99°C
Voltage 230 V
Electrical Requirements 230 V, 50/60 Hz, 700 W
Product Type Mastercycler Nexus Gradient
Height (Metric) 320 mm
Width (Metric) 250 mm
Depth (Metric) 410 mm
Frequency 50 to 60 Hz
Ramp Rate 3°C/s
Material Aluminum
Gradient Range 20°C