missing translation for 'onlineSavingsMsg'
Learn More

Origene Technologies MASP2 Monoclonal Antibody (OTI2F10), TrueMAB™, OriGene

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA812533S

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 17537014

  • 2453.92 NOK / 30µL
Estimated Shipment: 19-06-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MASP2 Monoclonal antibody specifically detects MASP2 in Human samples. It is validated for Western Blot
Specifications

Specifications

MASP2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O00187
Mouse
Protein A/G
RUO
10747
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI2F10
Unconjugated
MASP2
mannan binding lectin serine peptidase 2; mannan-binding; mannan-binding lectin serine peptidase 1 pseudogene 1; mannan-binding lectin serine peptidase 2; mannan-binding lectin serine peptidase 2 variant 1; mannan-binding lectin serine peptidase 2 variant 2; mannan-binding lectin serine peptidase 2 variant 3; mannan-binding lectin serine peptidase 2 variant 4; mannan-binding lectin serine protease 1 pseudogene 1; mannan-binding lectin serine protease 2; Mannan-binding lectin serine protease 2 A chain; Mannan-binding lectin serine protease 2 B chain; mannose binding lectin-associated serine protease-2; mannose-binding protein-associated serine protease 2; MAp19; MASP1P1; Masp2; MASP-2; MBL-associated plasma protein of 19 kD; MBL-associated serine protease 2; sb:eu714; small MBL-associated protein; sMAP; wu:fb62a09
Human recombinant protein fragment corresponding to amino acids 445-686 of human MASP2 (NP_006601) produced in E.coli.
30 μL
Primary
Human
Antibody
IgG2a
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers