missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MASP2 Monoclonal Antibody (OTI2F10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
2832.96 NOK - 6331.68 NOK
Specifications
| Antigen | MASP2 |
|---|---|
| Clone | OTI2F10 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17537014
|
Origene Technologies
TA812533S |
30 μL |
2832.96 NOK
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17517014
|
Origene Technologies
TA812533 |
100 μL |
6331.68 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MASP2 Monoclonal antibody specifically detects MASP2 in Human samples. It is validated for Western BlotSpecifications
| MASP2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| mannan binding lectin serine peptidase 2; mannan-binding; mannan-binding lectin serine peptidase 1 pseudogene 1; mannan-binding lectin serine peptidase 2; mannan-binding lectin serine peptidase 2 variant 1; mannan-binding lectin serine peptidase 2 variant 2; mannan-binding lectin serine peptidase 2 variant 3; mannan-binding lectin serine peptidase 2 variant 4; mannan-binding lectin serine protease 1 pseudogene 1; mannan-binding lectin serine protease 2; Mannan-binding lectin serine protease 2 A chain; Mannan-binding lectin serine protease 2 B chain; mannose binding lectin-associated serine protease-2; mannose-binding protein-associated serine protease 2; MAp19; MASP1P1; Masp2; MASP-2; MBL-associated plasma protein of 19 kD; MBL-associated serine protease 2; sb:eu714; small MBL-associated protein; sMAP; wu:fb62a09 | |
| Human recombinant protein fragment corresponding to amino acids 445-686 of human MASP2 (NP_006601) produced in E.coli. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| MASP2 |
| OTI2F10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O00187 | |
| 10747 | |
| IgG2a | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title