Learn More
Invitrogen™ Human MARK3, GST Tag Recombinant Protein
MARK3 is a member of a group of kinases known to regulate cell polarity via phosphorylation of microtubule-associated proteins (MAPs).
Brand: Invitrogen™ PV4819
Additional Details : Weight : 0.50000kg
Description
MARK3 is a member of a group of kinases known to regulate cell polarity via phosphorylation of microtubule-associated proteins (MAPs).Specifications
NP_002367.3 | |
4140 | |
10 μg | |
CAMLK Kinase Family, CAMKL Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.02% Trition X-100, 0.5 mM EDTA, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CAMK Group | |
Z'-LYTE™ | |
MAP⁄microtubule affinity-regulating kinase 3 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
108.9 kDa | |
MARK3 | |
Human | |
GST Tag | |
Biochemical phospho-specific activity (no antibody) | |
Dry Ice | |
MARK3 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |