Learn More
Invitrogen™ Human MARK3, GST Tag Recombinant Protein
MARK3 is a member of a group of kinases known to regulate cell polarity via phosphorylation of microtubule-associated proteins (MAPs).
5160.00 NOK - 112500.00 NOK
Specifications
Accession Number | NP_002367.3 |
---|---|
Molecular Weight (g/mol) | 108.9 kDa |
Gene Alias | MARK3 |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438693
|
Invitrogen™
PR8905A |
100μg |
17690.00 NOK
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15448763
|
Invitrogen™
PV4820 |
1mg |
112500.00 NOK
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10628703
|
Invitrogen™
PV4819 |
10 μg |
5160.00 NOK
Each |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MARK3 is a member of a group of kinases known to regulate cell polarity via phosphorylation of microtubule-associated proteins (MAPs).Specifications
NP_002367.3 | |
MARK3 | |
Recombinant | |
Baculovirus |
108.9 kDa | |
Human | |
GST Tag |