missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAPK9 (JNK2), His Tag Recombinant Protein
Human MAPK9 (JNK2), His Tag Recombinant Protein
Brand: Invitrogen™ PV4018
Additional Details : Weight : 0.00900kg
Description
- MAPK9 (JNK2) is a serine/threonine kinase which plays an important role in the MAPK pathway
Specifications
NP_002743 | |
52.2 kDa | |
MAPK9 (JNK2) | |
Human | |
His Tag | |
Ser/Thr Kinases: CMGC Group | |
Baculovirus | |
Frozen |
Kinase Assay | |
1 mg | |
MAP Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Full Length | |
Drug Development |