Learn More
Invitrogen™ Human MAPK9 (JNK2), His Tag Recombinant Protein
Human MAPK9 (JNK2), His Tag Recombinant Protein
5130.00 NOK - 69300.00 NOK
Specifications
Accession Number NP_002743
For Use With (Application) Kinase Assay
Molecular Weight (g/mol) 52.2 kDa
Gene Alias MAPK9 (JNK2)
KinaseFamily MAP Kinase Family
View More Specs
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11523477
View Documents Promotion Details
Invitrogen™
PV3620
10μg
5130.00 NOK
10µg
View alternative products
Save Up to 
»
In Stock
to see stock.
15438483
View Documents Promotion Details
Invitrogen™
PR6568B
100μg
16000.00 NOK
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-05-2024
to see stock.
10686013
View Documents Promotion Details
Invitrogen™
PV4018
1 mg
69300.00 NOK
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 03-05-2024
Description
  • MAPK9 (JNK2) is a serine⁄threonine kinase which plays an important role in the MAPK pathway.
  • Gibco™ MarrowMAX™ Bone Marrow Medium is a fully-supplemented medium developed for the short term culture of bone marrow and other hematopoietic cells for cytogenetic studies and in vitro diagnostic procedures.
Specifications
Specifications
NP_002743
52.2 kDa
MAP Kinase Family
Recombinant
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Baculovirus
Kinase Assay
MAPK9 (JNK2)
Human
His Tag
Dry Ice