Description
The MAP4K6 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_722549. The protein contains a C-terminal DDK tag. |
| For Use With (Application) | Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens MAP4K6, mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Research Category | MAP Kinase Signaling, Protein Kinase |
| Protein State | Native |
For Research Use Only