Learn More
Fisherbrand™ Manual Crimper, Steel, for Flip Top/Flip Off Seals or Flip Tear Up Seals
Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
For Flip Tear Up
For Flip Top/for Flip Off
Diameter (Metric) Cap:
13 mm
20 mm
28 mm
Unit Size:
Each
Specifications
Specifications
| Description | 13mm Crimper |
| Diameter (Metric) Cap | 13 mm |
| Material | Steel |
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | For Flip Top/for Flip Off |