Learn More
Fisherbrand™ Manual Crimper, Steel, for Aluminum/Steel caps
Product Code. 11839440 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Cap:
7.5 mm
8 mm
11 mm
13 mm
20 mm
28 mm
32 mm
Unit Size:
Each
Product Code. Diameter (Metric) Cap unitSize
11839440 28 mm Each
11815863 32 mm Each
11757577 11 mm Each
11748276 13 mm Each
11707567 8 mm Each
11888951 7.5 mm Each
11550525 20 mm Each
7 options
Product Code. 11839440
Only null left
Add to basket
Add to basket
Provide a reproducible, secure vial closure.
  • Crimper
  • Manual
  • Steel
  • Non-autoclavable
  • Adjustable
TRUSTED_SUSTAINABILITY
Specifications
Description 28mm Crimper
Diameter (Metric) Cap 28 mm
Material Steel