Learn More
Fisherbrand™ Manual Crimper, Steel, for Aluminum/Steel caps
Product Code. 11815863 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Cap:
7.5 mm
8 mm
11 mm
13 mm
20 mm
28 mm
32 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Cap unitSize
11815863 32 mm Each
11757577 11 mm Each
11748276 13 mm Each
11707567 8 mm Each
11888951 7.5 mm Each
11839440 28 mm Each
11550525 20 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11815863
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Provide a reproducible, secure vial closure.
  • Crimper
  • Manual
  • Steel
  • Non-autoclavable
  • Adjustable
TRUSTED_SUSTAINABILITY
Specifications
Description 32mm Crimper
Diameter (Metric) Cap 32 mm
Material Steel