Learn More
Cadence Science™ Malleable Stainless Steel Animal Feeding Needles
Product Code. 11948104
Change view
Click to view available options
Gauge:
18G
20G
Diameter (Metric) Ball:
1.9 mm
2.3 mm
Length (English):
1.5 in.
2 in.
3.1 in.
Unit Size:
Pack of 100
Product Code. Gauge Diameter (Metric) Ball Length (English) unitSize
11948104 18G 2.3 mm 2 in. Pack of 100
12393800 20G 1.9 mm 1.5 in. Pack of 100
16367555 18G 2.3 mm 3.1 in. Pack of 100
3 options
Product Code. 11948104
Brand: Cadence Science™ 9918
Designed to meet the special needs of pharmacologists, toxicologists, pathologists, and other biomedical researchers working with laboratory animals. Cadence Science™ Malleable Stainless Steel Animal Feeding Needles are sterile and disposable.
  • Type 304 stainless steel construction
  • Malleable
  • Sterile
  • Disposable
  • 100/box, 20 x 5/sterile pouch
Specifications
Product Type Animal Feeding Needle
Material Stainless Steel (304)
Disposable Single-use
For Use With (Application) Animal feeding
Gauge 18G
Sterility Sterile
Length (English) 2 in.
Diameter (Metric) Ball 2.3 mm