Learn More
BINDER Maintenance
Product Code. 30274305 Shop All BINDER Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers Series B, BD, BD-S, E, ED, ED-S
BINDER Chambers Series BF, FD, FD-S, FED, FP, M, FDL
BINDER Chambers Series CB-S, CB
BINDER Chambers Series CBF
BINDER Chambers Series KBF, KMF
BINDER Chambers Series KBF-S, KBF-S ECO
BINDER Chambers Series KT, KB, KB ECO
BINDER Chambers Series MK, MKT
BINDER Chambers Series MKF, MKFT
BINDER Chambers Series UF V
BINDER Chambers Series VD
BINDER Chambers Series VDL
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
30274305 BINDER Chambers Series KBF, KMF Each
30274312 BINDER Chambers Series B, BD, BD-S, E, ED, ED-S Each
30274300 BINDER Chambers Series CB-S, CB Each
30274282 BINDER Chambers Series MK, MKT Each
30274311 BINDER Chambers Series UF V Each
30274313 BINDER Chambers Series BF, FD, FD-S, FED, FP, M, FDL Each
30274299 BINDER Chambers Series CBF Each
30274281 BINDER Chambers Series VD Each
30274273 BINDER Chambers Series KBF-S, KBF-S ECO Each
30274284 BINDER Chambers Series KT, KB, KB ECO Each
30274275 BINDER Chambers Series MKF, MKFT Each
30274294 BINDER Chambers Series VDL Each
12 options
Product Code. 30274305
Brand: BINDER DL200613
One-off maintenance service in accordance with maintenance schedule.
One-off maintenance service in accordance with maintenance schedule.
  • Visual inspection of mechanical and electrical components, testing of all key functions.
  • Calibration of a test temperature in the center of usable space as specified by the user
  • Without certificate
Specifications
For Use With (Equipment) BINDER Chambers Series KBF, KMF
Product Type Preventive Maintenance