Description
When tubes are placed in the rack, the magnetic beads are attracted to the magnet within a few seconds. Tubes can be locked in position and the rack inverted, which allows easy removal of waste solutions between steps in the protocol, while the magnetic beads remain in the tube.
The rack consists of anodized aluminum housing with a detachable plastic magnetic bar containing six neodymium magnets.
- Rapid and simple processing for Mag Sepharose™ purification and enrichment applications
- No need for centrifugation
- Vary the amount of beads to meet specific requirements
Specifications
Specifications
| For Use With (Application) | For use with products containing magnetic beads |
| Product Type | MagRack 6 |