Learn More
Invitrogen™ MagniSort™ Streptavidin Positive Selection Beads
Product Code. 15582567
Click to view available options
Quantity:
200 Tests
25 Tests
Unit Size:
200 tests
25 tests
Contains Sodium azide
Product Code. 15582567
Brand: Invitrogen™ MSPB600371
Contains Sodium azide
Designed for the magnetic separation of cells by positive selection. Desired cells are bound by biotinylated antibody and then streptavidin-coated magnetic beads. When placed in a magnetic field, desired cells are held in place and undesired cells can be separated by decanting.
  • Clone and concentration of biotinylated antibody for desired cell labeling, as well as concentration of MagniSort™ Streptavidin Positive Selection Beads must be determined empirically
  • Please refer to the protocol for details on how to optimize both the antibody and MagniSort™ Streptavidin Positive Selection Beads
  • The MagniSort™ Streptavidin Positive Selection Beads may also be used to deplete undesired cells using a single antibody
  • For depletion of cells using multiple antibodies, we recommend the MagniSort™ Streptavidin Negative Selection Beads (cat. MSNB-6002) and protocol
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 Tests
Form Liquid
Ligand Streptavidin
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin