Learn More
Invitrogen™ MagniSort™ Streptavidin Positive Selection Beads
Click to view available options
Quantity:
200 Tests
25 Tests
Unit Size:
200 tests
25 tests
Description
- Clone and concentration of biotinylated antibody for desired cell labeling, as well as concentration of MagniSort™ Streptavidin Positive Selection Beads must be determined empirically
- Please refer to the protocol for details on how to optimize both the antibody and MagniSort™ Streptavidin Positive Selection Beads
- The MagniSort™ Streptavidin Positive Selection Beads may also be used to deplete undesired cells using a single antibody
- For depletion of cells using multiple antibodies, we recommend the MagniSort™ Streptavidin Negative Selection Beads (cat. MSNB-6002) and protocol
Specifications
Specifications
| Quantity | 200 Tests |
| Form | Liquid |
| Ligand | Streptavidin |
| Product Type | Streptavidin Conjugate (Fluorescent) |
| Target Molecule | Biotin |