missing translation for 'onlineSavingsMsg'
Learn More

SP Bel-Art™ Magnetic Stirring Bars Spinbar™

Heavy duty bars for use in stirring large volumes of fluid utilizing large magnetic stirring machines such as the Cool Stir Large Volume Stirrer, H37028-0001 or other large machines

Brand:  SP Bel-Art™ F37118-1600

Additional Details : Weight : 4.36020kg

 View more versions of this product

Product Code. 16448495

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Will fit into vessels with a minimum neck diameter of 32 mm
  • Multi-faceted surface adds turbulence relative to similar size smooth cylindrical stirring bars
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE Coating; Superior chemical compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Specifications

Specifications

28 mm
1 Ea.
160 mm
Giant Polygon
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers