missing translation for 'onlineSavingsMsg'
Learn More

SP Bel-Art™ Magnetic Stirring Bars Spinbar™

Heavy duty bars for use in stirring large volumes of fluid utilizing large magnetic stirring machines such as the Cool Stir Large Volume Stirrer, H37028-0001 or other large machines

920.00 NOK - 935.00 NOK

Specifications

Diameter (Metric) 28 mm
Quantity 1 Ea.
Shape Giant Polygon
View More Specs

Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
16438495
View Documents Promotion Details
SP Bel-Art™
F37118-1100
110 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16448495
View Documents Promotion Details
SP Bel-Art™
F37118-1600
160 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Will fit into vessels with a minimum neck diameter of 32 mm
  • Multi-faceted surface adds turbulence relative to similar size smooth cylindrical stirring bars
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE Coating; Superior chemical compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Specifications

Specifications

28 mm
1 Ea.
Giant Polygon
Videos
SDS
Documents

Documents

Product Certifications