Learn More
Gilson™ MACROMAN™ Charging Desk Stand for MACROMAN Pipette Controller
Product Code. 15881727 Shop All Gilson Products
Change view
Click to view available options
:
Desk stand; accessory
Product Code.
15881727 Desk stand; accessory
1 options
Product Code. 15881727
Brand: Gilson™ F107200
Only null left
Add to basket
Add to basket
An accessory charging desk stand for use with the MACROMAN™ pipette controller.
  • Supplied individually
  • Black in color
  • Accessory for MACROMAN pipette controllers
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Type Desktop Charger Stand
For Use With (Application) Charging and storage
For Use With (Equipment) MACROMAN™ Pipette Controller
Quantity 1