missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYVE-1 Antibody (ALY7) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 15 publications
3140.00 NOK - 4760.00 NOK
Tekniske data
| Antigen | LYVE-1 |
|---|---|
| Clone | ALY7 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry <= 0.125 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 2.5 ug/ml |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|
18084415
|
Novus Biologicals
NBP1-43411-0.025MG |
0.025 mg |
3140.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18785353
|
Novus Biologicals
NBP1-43411-0.1mg |
0.1 mg |
4760.00 NOK
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
LYVE-1 Monoclonal specifically detects LYVE-1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Tekniske data
| LYVE-1 | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Mouse | |
| Q8BHC0 | |
| 10894 | |
| The immunogen for this antibody is mouse LYVE1. | |
| Primary | |
| Store at 4C. Do not freeze. |
| ALY7 | |
| Flow Cytometry <= 0.125 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 2.5 ug/ml | |
| Monoclonal | |
| Rat | |
| Endosome Markers, Endothelial Cell Markers, Immunology | |
| PBS, pH 7.2 with 0.09% Sodium Azide | |
| cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE-1XLKD1 | |
| LYVE1 | |
| IgG1 | |
| Affinity Purified | |
| Human reactivity reported in scientific literature (PMID: 23975155) |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel