missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYVE-1 Antibody (ALY7) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 15 publications
Brand: Novus Biologicals NBP1-43411-0.025MG
This item is not returnable.
View return policy
Description
LYVE-1 Monoclonal specifically detects LYVE-1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| LYVE-1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry <= 0.125 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 2.5 ug/ml | |
| Q8BHC0 | |
| LYVE1 | |
| The immunogen for this antibody is mouse LYVE1. | |
| 0.025 mg | |
| Endosome Markers, Endothelial Cell Markers, Immunology | |
| 10894 | |
| Mouse | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| ALY7 | |
| Unconjugated | |
| PBS, pH 7.2 with 0.09% Sodium Azide | |
| cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE-1XLKD1 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human reactivity reported in scientific literature (PMID: 23975155) | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction