Learn More
Cytiva LV-4 Manual Valve
May be inserted into the flow path without causing unnecessary mixing or sample dilution
Brand:  Cytiva 19-0017-01
Additional Details : Weight : 0.07600kg
Product Code. 12492300
  • 5829.00 NOK / Each
Estimated Shipment: 12-07-2024
Description
Two low pressure, manual valve series are available including the economical LV series designed for C column systems (see C Columns) and for other applications at pressures up to 3 bar (0.3 MPa, 42 psi).
  • A range of manual and automated valves to meet a variety of performance requirements.
  • Designed for biocompatibility and years of reliable service, each may be inserted into the flow path without causing unnecessary mixing or sample dilution.
Specifications
Specifications
3 bar
1 mm