Learn More
Molecular Probes™ Lucifer Yellow Biocytin, Potassium Salt (Biocytin Lucifer Yellow)
lucifer yellow biocytin, potassium salt (biocytin lucifer yellow)
Brand: Molecular Probes™ L6950
Additional Details : Weight : 0.00450kg
Description
The water-soluble neuronal tracer lucifer yellow biocytin can be detected in fixed tissues by using avidin or streptavidin probes. However, lucifer yellow biocytin can also be observed directly in fixed or unfixed samples because the attached lucifer yellow dye produces yellow fluorescence (absorption/emission maxima ∼428/536 nm).Specifications
Lucifer Yellow Biocytin | |
Store at room temperature and protect from light. | |
Room Temperature | |
Yellow |
5 mg | |
Biocytins | |
Water | |
Other Labels or Dyes |
For Research Use Only. Not for use in diagnostic procedures.