Learn More
Molecular Probes™ Lucifer Yellow Biocytin, Potassium Salt (Biocytin Lucifer Yellow)
lucifer yellow biocytin, potassium salt (biocytin lucifer yellow)
Brand:  Molecular Probes™ L6950
Additional Details : Weight : 0.00450kg
Product Code. 11564976
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
The water-soluble neuronal tracer lucifer yellow biocytin can be detected in fixed tissues by using avidin or streptavidin probes. However, lucifer yellow biocytin can also be observed directly in fixed or unfixed samples because the attached lucifer yellow dye produces yellow fluorescence (absorption/emission maxima ∼428/536 nm).
Specifications
Specifications
Lucifer Yellow Biocytin
Store at room temperature and protect from light.
Room Temperature
Yellow
5 mg
Biocytins
Water
Other Labels or Dyes