Learn More
WTW™ LTA 1 Special Conductivity Cell
Click to view available options
Cell Constant:
1 cm-1
Unit Size:
Each
Specifications
Specifications
| Cell Constant | 1 cm-1 |
| Type | Special Conductivity Cell |
| Includes | 2-electrode, adapter cable K/LTA |
| Model | LTA 1 |