Learn More
WTW™ LR 325/001 Pure Water Conductivity Measuring Cells
GREENER_CHOICE
Product Code. 10490632
Change view
Click to view available options
Length (Metric) Cable:
1.5 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Cable Unit Size
10490632 1.5 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10490632 Supplier WTW™ Supplier No. 301962
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
In Stock
Two electrode measuring cells with concentric electrodes of stainless steel for measurements in pure water.
  • Precise measured values thanks to concentric elecrodes without edge fields
  • Includes flow vessels to prevent incorrect results due to carbon dioxide
  • No polorization effects thanks to stainless steel design
TRUSTED_SUSTAINABILITY
Specifications
Cell Constant 0.01 cm-1
For Use With (Application) Measuring the intrinsic conductivity of purified water in the pharmaceutical industry and the production of semiconductors
Temperature (Metric) Range 0°C to 100°C
Type Standard Conductivity Cell
Diameter (Metric) Body 20 mm
Length (Metric) Cable 1.5 m
Material Steel, V4A
Includes Temperature sensor, 1.5m Length cable with waterproof plug
Model LR 325/001