Learn More
Fisherbrand™ LPG Gas Security Bunsen Burners with Tétine Connector
Product Code. 11799777 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Gas Type:
Butane, LPG, Propane
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gas Type Unit Size
11799777 Butane, LPG, Propane Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11799777
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
Supplied with a flame failure device that shuts off the gas supply to the burner within approximately 20 seconds of the flame being extinguished.
  • Safety features all contained within the burner
  • For use with natural gas/methane or butane/propane
  • 13mm diameter opening
  • Chrome steel burner, cast iron base
  • When igniting the flame, keep the gas inlet valve open for 3 to 5 seconds
Specifications
Product Type Safety Bunsen Burner
Gas Type Butane, LPG, Propane
Diameter (Metric) 13 mm
For Use With (Application) Tétine Connector
Height (Metric) 165 mm
Includes T-Tine Connector
No. per Pack 1 Ea.