Learn More
Eppendorf™ Locking Clip
Product Code. 15234326 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
12-channel pipette
8-channel pipette
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15234326 12-channel pipette Each
15204326 8-channel pipette Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15234326 Supplier Eppendorf™ Supplier No. 3122613002
  • ONLINE EXCLUSIVE PRICE
    103.00 NOK
    100.00 NOK / Pack of 1
    Save 3.00 NOK
    2% Off
In Stock
Prevent spring action of tip cones with these locking clips
  • Accessory for Eppendorf Xplorer™/Xplorer plus
  • Locking clip
  • Available for 8- and 12-channel pipettes
TRUSTED_SUSTAINABILITY
Specifications
Type Locking Clip
For Use With (Application) Preventing spring action of tip cones
For Use With (Equipment) 12-channel pipette
Quantity 1 Each