Learn More
Novus Biologicals™ LOC554223 293T Cell Transient Overexpression Lysate
Product Code. 18598433 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18598433 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18598433 Supplier Novus Biologicals™ Supplier No. H00554223T01
 Restricted Item
Estimated Shipment: 15-10-2026
Specifications
Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Target LOC554223
Synonym hypothetical LOC554223
Gene Symbol LOC554223