Learn More
Brannan™ LO-tox™ Lab Thermometer with 3-points UKAS Calibration
Product Code. 17843078
Click to view available options
Temperature Range (Metric):
-1°C to 101°C
-1°C to 51°C
-10°C to +110°C
-10°C to +210°C
-10°C to +260°C
-10°C to +300°C
-10°C to +360°C
-10°C to +50°C
-20°C to +110°C
-20°C to +150°C
-35°C to +50°C
Subdivision:
0.1°C
0.2°C
0.5°C
1°C
2°C
Immersion Type:
Partial Immersion
Total Immersion
Length (Metric):
15.5 cm
30.5 cm
40.5 cm
Unit Size:
Each
Product Code. 17843078
Brand: Brannan™ 44/850/7 + 49/503/0
LO-tox™ lab thermometer complete with calibration certificate.
  • LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
  • Safe non-mercury filling
  • Supplied with calibration certificate
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK
Specifications
Calibration Points 0°C, 20°C, 45°C
Certifications/Compliance ISO 17025 UKAS
Immersion Type Total Immersion
Length (Metric) 15.5 cm
Material Glass
Thermometer Fill Blue Spirit
Max. Temperature (English) 122°F
Max. Temperature (Metric) +50°C
Min. Temperature (English) 14°F
Min. Temperature (Metric) -10°C
Temperature Range (English) 14°F to 122°F
Temperature Range (Metric) -10°C to +50°C
Accuracy ± 1.0°C at 35°C
Subdivision 1°C
Width (Metric) 1.2 cm