Learn More
Brannan™ LO-tox™ Lab Thermometer with 3-points UKAS Calibration
Click to view available options
Temperature Range (Metric):
-1°C to 101°C
-1°C to 51°C
-10°C to +110°C
-10°C to +210°C
-10°C to +260°C
-10°C to +300°C
-10°C to +360°C
-10°C to +50°C
-20°C to +110°C
-20°C to +150°C
-35°C to +50°C
Subdivision:
0.1°C
0.2°C
0.5°C
1°C
2°C
Immersion Type:
Partial Immersion
Total Immersion
Length (Metric):
15.5 cm
30.5 cm
40.5 cm
Unit Size:
Each
Description
- LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
- Safe non-mercury filling
- Supplied with calibration certificate
- Non-roll end cap
- Supplied as standard in protective individual, resealable telescopic tube
- Made in UK
Specifications
Specifications
| Calibration Points | 0°C, 100°C, 200°C |
| Certifications/Compliance | ISO 17025 UKAS |
| Immersion Type | Partial Immersion |
| Length (Metric) | 40.5 cm |
| Material | Glass |
| Thermometer Fill | Blue Spirit |
| Max. Temperature (English) | 410°F |
| Max. Temperature (Metric) | +210°C |
| Min. Temperature (English) | 14°F |
| Min. Temperature (Metric) | -10°C |
| Show More |