missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ LMO2 (Human) Recombinant Protein

Human LMO2 full-length ORF ( AAH34041, 1 a.a. - 158 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00004005-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16137431

  • 4115.00 NOK / 10µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MSSAIERKSLDPSEEPVDEVLQIPPSLLTCGGCQQNIGDRYFLKAIDQYWHEDCLSCDLCGCRLGEVGRRLYYKLGRKLCRRDYLRPFGQDGLCASCDKRIRAYEMTMRVKDKVYHLECFKCAACQKHFCVGDRYLLINSDIVCEQDIYEWTKINGMI
Specifications

Specifications

AAH34041
LIM domain only 2 (rhombotin-like 1)
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LMO2
GST
4005
Wheat germ expression system
10 μg
RBTN2, RBTNL1, RHOM2, TTG2
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers