missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ LMO2 (Human) Recombinant Protein
Human LMO2 full-length ORF ( AAH34041, 1 a.a. - 158 a.a.) recombinant protein with GST-tag at N-terminal.
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH34041 |
---|---|
Gene ID (Entrez) | 4005 |
Name | LIM domain only 2 (rhombotin-like 1) |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16137431
|
Abnova™
H00004005-P01.10UG |
10 μg |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16147431
|
Abnova™
H00004005-P01.25UG |
25 μg |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MSSAIERKSLDPSEEPVDEVLQIPPSLLTCGGCQQNIGDRYFLKAIDQYWHEDCLSCDLCGCRLGEVGRRLYYKLGRKLCRRDYLRPFGQDGLCASCDKRIRAYEMTMRVKDKVYHLECFKCAACQKHFCVGDRYLLINSDIVCEQDIYEWTKINGMI
Specifications
AAH34041 | |
LIM domain only 2 (rhombotin-like 1) | |
125% SDS-PAGE Stained with Coomassie Blue | |
RBTN2, RBTNL1, RHOM2, TTG2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
4005 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LMO2 | |
GST |