missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ LMO2 (Human) Recombinant Protein
Human LMO2 full-length ORF ( AAH34041, 1 a.a. - 158 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004005-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSSAIERKSLDPSEEPVDEVLQIPPSLLTCGGCQQNIGDRYFLKAIDQYWHEDCLSCDLCGCRLGEVGRRLYYKLGRKLCRRDYLRPFGQDGLCASCDKRIRAYEMTMRVKDKVYHLECFKCAACQKHFCVGDRYLLINSDIVCEQDIYEWTKINGMI
Specifications
AAH34041 | |
LIM domain only 2 (rhombotin-like 1) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LMO2 | |
GST |
4005 | |
Wheat germ expression system | |
25 μg | |
RBTN2, RBTNL1, RHOM2, TTG2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |