Learn More
Novus Biologicals™ Lightning-Link (R) Rapid FluoProbes647H Antibody Labeling Kit
Shop All Bio Techne ProductsDescription
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 30 μg |
| Sufficient For | 3 x 20 μg Reactions |
| Product Type | Rapid FluoProbes647H Antibody Labeling Kit |