missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid FluoProbes647H Antibody Labeling Kit
Shop All Bio Techne Products
Click to view available options
Sufficient For:
1 x 2 mg Reaction
3 x 20 μg Reactions
3 x 200 μg Reactions
Unit Size:
1 set
1mg
30µg
Description
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 30 μg |
| Sufficient For | 3 x 200 μg Reactions |
| Product Type | Rapid FluoProbes647H Antibody Labeling Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction