Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Atto390 Antibody Labeling Kit
Product Code. 18033396 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. 18033396
Brand: Novus Biologicals™ 3490005
Antibody labeling kit
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
  • Lightning-Link™ Rapid mix
  • 1 vial of LL Rapid Quencher reagent
  • 1 vial of LL Rapid Modifier reagent
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 μg
Product Type Antibody Labeling Kit
Content And Storage -20°C