Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Alexa Fluor 700 Antibody Labeling Kit
Product Code. 18053736 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
Unit Size:
1 set
100µg
1mg
Product Code. Quantity unitSize
18053736 1 mg 1mg
18045336 100 μg 100µg
18066124 300 μg 1 set
18033484 30 μg 1 set
4 options
Product Code. 18053736
Brand: Novus Biologicals™ 3370015
Antibody labeling kit
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
  • Lightning-Link™ Rapid mix
  • 1 vial of LL Rapid Quencher reagent
  • 1 vial of LL Rapid Modifier reagent
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mg
Product Type Antibody Labeling Kit
Content And Storage -20°C