Learn More
Purolite™ Lifetech™ ECR Resins for Enzyme Immobilization, 300 to 710 μm
Product Code. 17049383
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Pore Size:
1200 to 1800 Å
220 to 340 Å
300 to 600 Å
400 to 650 Å
600 to 1200 Å
Unit Size:
1kg
250g
50g
Product Code. Quantity Pore Size unitSize
17049383 250 g 400 to 650 Å 250g
17009343 1 kg 1200 to 1800 Å 1kg
17009373 1 kg 300 to 600 Å 1kg
17019323 250 g 220 to 340 Å 250g
17019383 250 g 1200 to 1800 Å 250g
17029323 1 kg 220 to 340 Å 1kg
17029383 1 kg 1200 to 1800 Å 1kg
17039333 250 g 600 to 1200 Å 250g
17039363 250 g 1200 to 1800 Å 250g
17049333 1 kg 600 to 1200 Å 1kg
17049353 250 g 600 to 1200 Å 250g
17049363 1 kg 1200 to 1800 Å 1kg
17059353 1 kg 600 to 1200 Å 1kg
17059373 250 g 600 to 1200 Å 250g
17059383 1 kg 400 to 650 Å 1kg
17069373 1 kg 600 to 1200 Å 1kg
17079313 50 g - 50g
17079323 250 g 300 to 600 Å 250g
17079353 250 g 600 to 1200 Å 250g
17089313 250 g - 250g
17089323 1 kg 300 to 600 Å 1kg
17089343 250 g 300 to 600 Å 250g
17089353 1 kg 600 to 1200 Å 1kg
17099313 1 kg - 1kg
17099333 250 g 1200 to 1800 Å 250g
17099343 1 kg 300 to 600 Å 1kg
17099363 250 g 300 to 600 Å 250g
27 options
Product Code. 17049383
Brand: Purolite™ LS01415192
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Lifetech™ ECR resins are robust polymeric resins designed for research and industrial immobilization of enzymes. Available for covalent, ionic, adsorption and affinity immobilization.
  • Enzyme immobilization by adsorption, covalent, IEX and affinity
  • High mechanical stability
  • High porosity
  • Easily dried after immobilization
  • Suitable for batch and column configuration
  • Optimal for use in organic solvent/water free systems
  • Compliant with ResAP(2004) 3
  • Halal, Kosher and TSE/BSE/GMO free
Specifications
pH Range 1 to 14
Pore Size 400 to 650 Å
Content And Storage 2°C to 20°C
Particle Size 300 to 710 μm
Moisture 57 to 68%
Functionality/Interaction Type Octadecyl
Form Wet
Product Line Lifetech
Quantity 250 g
Type Methacrylate (ECR Resin)
Format White to Off-White Spherical Beads