Learn More
Purolite™ Lifetech™ ECR Resins for Enzyme Immobilization, 300 to 710 μm
Click to view available options
Quantity:
50 g
250 g
1 kg
Pore Size:
1200 to 1800 Å
220 to 340 Å
300 to 600 Å
400 to 650 Å
600 to 1200 Å
Unit Size:
1kg
250g
50g
Description
- Enzyme immobilization by adsorption, covalent, IEX and affinity
- High mechanical stability
- High porosity
- Easily dried after immobilization
- Suitable for batch and column configuration
- Optimal for use in organic solvent/water free systems
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifications
Specifications
| pH Range | 1 to 14 |
| Pore Size | 220 to 340 Å |
| Content And Storage | 2°C to 20°C |
| Particle Size | 300 to 710 μm |
| Moisture | 57 to 68% |
| Form | Wet |
| Product Line | Lifetech |
| Quantity | 250 g |
| Type | Methacrylate (ECR Resin) |
| Format | White to Off-White Spherical Beads |