Learn More
Fisherbrand™ Lids for Desiccator with nz 24/29
Product Code. 15524635 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Outer:
153 mm
215 mm
270 mm
320 mm
380 mm
Height (Metric):
64 mm
89 mm
97 mm
111 mm
136 mm
Description:
Type: 100 mm
Type: 150 mm
Type: 200 mm
Type: 250 mm
Type: 300 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Height (Metric) Description Unit Size
15524635 270 mm 97 mm Type: 200 mm Each
15574635 320 mm 111 mm Type: 250 mm Each
15574625 215 mm 89 mm Type: 150 mm Each
15524645 380 mm 136 mm Type: 300 mm Each
15524625 153 mm 64 mm Type: 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15524635
  • ONLINE EXCLUSIVE PRICE
    1425.00 NOK
    1244.51 NOK / Pack of 1
    Save 180.49 NOK
    12% Off
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
For use with desiccators with nz 24/29
  • For use with desiccators with nz 24/29
Certifications
ISO 13 130, DIN 12 491
Specifications
Diameter (Metric) Outer 270 mm
Description Type: 200 mm
For Use With (Equipment) Desiccator
Height (Metric) 97 mm
Product Type Desiccator Lid