Learn More
Fisherbrand™ Lids for Desiccator with nz 24/29
Product Code. 15524625 Shop All Fisherbrand Products
missing translation for 'orderingAttributeHoverText'
Diameter (Metric) Outer:
153 mm
215 mm
270 mm
320 mm
380 mm
Height (Metric):
64 mm
89 mm
97 mm
111 mm
136 mm
Description:
Type: 100 mm
Type: 150 mm
Type: 200 mm
Type: 250 mm
Type: 300 mm
missing translation for 'unitSize'
Each
Product Code. 15524625
Only null left
Add to basket
Add to basket
For use with desiccators with nz 24/29
  • For use with desiccators with nz 24/29
Certifications
ISO 13 130, DIN 12 491
Specifications
Diameter (Metric) Outer 153 mm
Product Type Lid
Description Type: 100 mm
For Use With (Equipment) Desiccator
Height (Metric) 64 mm