Learn More
Fisherbrand™ Lid for Desiccator with Glass Knob
Product Code. 15564635 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Outer:
153 mm
215 mm
270 mm
320 mm
380 mm
Height (Metric):
78 mm
103 mm
108 mm
125 mm
150 mm
Description:
Type: 100 mm
Type: 150 mm
Type: 200 mm
Type: 250 mm
Type: 300 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Height (Metric) Description Unit Size
15564635 320 mm 125 mm Type: 250 mm Each
15564625 215 mm 103 mm Type: 150 mm Each
15514645 380 mm 150 mm Type: 300 mm Each
15514635 270 mm 108 mm Type: 200 mm Each
15514625 153 mm 78 mm Type: 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15564635
  • ONLINE EXCLUSIVE PRICE
    1330.00 NOK
    1161.38 NOK / Pack of 1
    Save 168.62 NOK
    12% Off
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
For use with desiccators with glass knob
  • For use with desiccators with glass knob
Certifications
ISO 13 130, DIN 12 491
Specifications
Diameter (Metric) Outer 320 mm
Description Type: 250 mm
For Use With (Equipment) Desiccator with glass knob
Height (Metric) 125 mm
Product Type Desiccator Lid