Learn More
Elma™ Level Sets
Product Code. 17891327
Change view
Click to view available options
Model:
L
M
S
XL
Unit Size:
1 set
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
17891327 S 1 set
17841347 M 1 set
17811337 L Each
17831357 XL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17891327 Supplier Elma™ Supplier No. 1125225
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Modular basket system for the Elmasonic P, Elmasonic Select, and EASY.
  • Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
  • Ergonomic handles
  • Autoclavable
  • Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
  • Square openings in the side walls for mounting silicone holders
Specifications
Type Level Set
For Use With (Equipment) Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
Model S