Learn More
Novus Biologicals™ LDLR Antibody Pair [HRP]
Product Code. 18295266 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Product Code. 18295266
Brand: Novus Biologicals™ NBP27954015PLATES
Mouse Monoclonal Antibody Pair
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human LDLR.
TRUSTED_SUSTAINABILITY
Spécification
Assay Range 93.75 pg/mL
Assay Sensitivity 93.75-6000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human LDLR.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Gene ID (Entrez) 3949
Gene Symbol LDLR
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 88 ng of recombinant Human LDLR.
  • Capture Antibody: 1.5 mg/mL of mouse anti-Human LDLR monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human LDLR monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
    Storage Requirements Storage is content dependent.
    Target LDLR
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 1 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 6000 pg/mL is recommended.
    Afficher plus Afficher moins
    Correction du contenu d'un produit
    Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
    Nom du produit
    En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
    Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis