Learn More
MP Biomedicals™ LB Medium, Large Capsules
Culture media used for the growth of bacteria.
742.00 NOK - 41085.60 NOK
Specifications
Certifications/Compliance | Molecular Biology Certified™ |
---|---|
Content And Storage | 15°C to 30°C |
Format | Capsule |
Formulation | Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L |
For Use With (Application) | For growth of bacteria |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11363229
|
MP Biomedicals™
113002036 |
1000 g |
3259.50 NOK
1kg |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
11393229
|
MP Biomedicals™
113002046 |
5000 g |
14182.80 NOK
5kg |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11393219
|
MP Biomedicals™
113002016 |
227 g |
742.00 NOK
227g |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11333229
|
MP Biomedicals™
113002026 |
454 g |
1542.30 NOK
454g |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
11323239
|
MP Biomedicals™
113002056 |
15 kg |
41085.60 NOK
15kg |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
Description
- Formulations for the E. coli growth media contain 10g of tryptone and 5g of yeast extract per liter, 10g NaCl per liter
Specifications
Molecular Biology Certified™ | |
Capsule | |
For growth of bacteria |
15°C to 30°C | |
Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L |